Cyclo(-D-Tyr-Arg-Gly-Asp-Cys(carboxymethyl)-OH) sulfoxide
Supplier: BACHEM AMERICAS INC
4020214.0001
4020214.0005
N-1365.0001BAEA
583.42
CAD
N-1365.0001BA
N-1365.0005BA
Cyclo(-D-Tyr-Arg-Gly-Asp-Cys(carboxymethyl)-OH) sulfoxide
Proteins and Peptides
This sulfur-bridged cyclopeptide is a potent inhibitor of the binding of glycoprotein IIb/IIIa to fibrinogen with an IC₅₀ of 1.5 nM. Furthermore, it is a highly potent inhibitor of fibrinogen-mediated platelet aggregation (IC₅₀ = 0.15 µM). Its antithrombotic activity in vivo may render this compound useful in the treatment of arterial thromboembolic diseases.