Ac-Tyr-Val-Ala-Asp-chloromethylketone
Supplier: BACHEM AMERICAS INC
4018838.0005 4018838.0025
N-1330.0005BAEA 434.83 CAD
N-1330.0005BA N-1330.0025BA
Ac-Tyr-Val-Ala-Asp-chloromethylketone
Proteins and Peptides
See also the Asp derivatives N-1425 and N-1430, and Z-YVAD-FMK (N-1595).
Cell-permeable, irreversible inhibitor of caspase-1 (ICE). N.Rouquet et al. showed that Ac-YVAD-CMK is a potent therapeutic agent against in vivo liver apoptosis in mice.