H-Glu-Gly-Arg-chloromethylketone Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4018214.0005
4018214.0025
N-1325.0005BAEA
903.62
CAD
N-1325.0005BA
N-1325.0025BA
H-Glu-Gly-Arg-chloromethylketone Trifluoroacetate
Proteins and Peptides
Peptide and amino acid chloromethyl ketones (CMK's) are irreversible inhibitors of cysteine proteases, but they also react with serine proteases. For our amino acid CMK's (e.g. TPCK, TLCK) see the corresponding product family in the 'Amino acid' section.
GGACK (EGR-CMK), inhibitor of the u-plasminogen activator.