MeOSuc-Ala-Ala-Pro-Ala-chloromethylketone
Supplier: BACHEM AMERICAS INC
4012071.0025 4012071.0100
N-1280.0025BAEA 903.62 CAD
N-1280.0025BA N-1280.0100BA
MeOSuc-Ala-Ala-Pro-Ala-chloromethylketone
Proteins and Peptides
See also H-8150 and N-1405.
MeOSuc-AAPA-CMK, inhibitor of human neutrophil elastase. The structure of the enzyme-inhibitor complex at 1.84 Å resolution reveals crosslinking of the catalytic residues His-57 and Ser-195 by the peptidyl chloromethylketone.