Acetyl-Pepstatin
Supplier: BACHEM AMERICAS INC
4015928.0001
4015928.0005
N-1250.0001BAEA
317.42
CAD
N-1250.0001BA
N-1250.0005BA
Acetyl-Pepstatin
Proteins and Peptides
Acetyl-pepstatin is reported to be an effective inhibitor of HIV-1 proteinase (Ki = 20 nM at pH 4.7) and of HIV-2 proteinase (Ki = 5 nM at pH 4.7). Ac-Val-Val-Sta-Ala-Sta also inhibits the P. falciparum aspartyl protease plasmepsin II, IC₅₀ 0.6 nM.