H-D-Phe-Phe-Arg-chloromethylketone Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4014271.0025 4014271.0100
N-1215.0025BAEA 1884.69 CAD
N-1215.0025BA N-1215.0100BA
H-D-Phe-Phe-Arg-chloromethylketone Trifluoroacetate
Proteins and Peptides
See also H-1765, H-6560, and N-1000.
Kallikrein inhibitor which is even effective in preventing apolipoprotein B-100 proteolysis in dextran sulfate (DS)-precipitated human plasma low density lipoproteins (LDL). D-FFR-CMK also inhibits gingipain R.