H-Val-Trp-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-((S)-2-Amino-3-methylbutanamido)-3-(1H-indol-3-yl)propanoic acid

4008196.0250 4008196.1000
N-1170.0250BAEA 413.58 CAD
N-1170.0250BA N-1170.1000BA
H-Val-Trp-OH
H-Val-Trp-OH

Sequence: H-Val-Trp-OH


These short peptides act as angiotensin I-converting enzyme (ACE) inhibitors. See also Phe-Tyr (G-2955) and the subfamilies 'Bradykinin-Potentiating Peptide (BPP)' and 'Angiotensin I-Converting Enzyme (ACE, ACE2) Inhibitors'.


The dipeptide Val-Trp is a potent, competitive dipeptide angiotensin I-converting enzyme inhibitor with a Ki of 0.3 µM.

Formula: C₁₆H₂₁N₃O₃
MW: 303.36 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037947
CAS Number: 24587-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR