Methyl L-tryptophanate hydrochloride
Supplier: BACHEM AMERICAS INC
4000133.0025
4000133.0100
N-1165.0025BAEA
490.51
CAD
N-1165.0025BA
N-1165.0100BA
Methyl L-tryptophanate hydrochloride
Methyl L-tryptophanate hydrochloride
Sequence: H-Trp-OMe · HCl
A competitive inhibitor of dexamethasone binding to the glucocorticoid receptor in HTC cells.
Formula:
C₁₂H₁₄N₂O₂·HCl Storage Temperature: Freezer |
MDL Number:
MFCD00066134 CAS Number: 7524-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...