N-(p-Toluenesulfonyl)-L-phenylalanylchloromethylketone
Supplier: BACHEM AMERICAS INC
Synonyms:
L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, (S)-N-(4-Chloro-3-oxo-1-phenyl-2-butyl)-p-toluenesulfonamide, N-Tosyl-L-phenylalanyl Chloromethyl Ketone, (S)-N-(4-Chloro-3-oxo-1-phenyl-2-butyl)-p-toluenesulphonamide, TPCK
4000836.0001
4000836.0005
N-1160.0001BAEA
217.06
CAD
N-1160.0001BA
N-1160.0005BA
N-(p-Toluenesulfonyl)-L-phenylalanylchloromethylketone
N-(p-Toluenesulfonyl)-L-phenylalanylchloromethylketone
Sequence: Tos-Phe-chloromethylketone
Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
See also Z-Phe-OH (C-2310) and Z-Phe-ONp (C-2345).
Synonym(s): TPCK
Formula:
C₁₇H₁₈ClNO₃S Storage Temperature: Freezer |
MDL Number:
MFCD00000935 CAS Number: 402-71-1 EINECS: 206-954-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...