(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride

Supplier: BACHEM AMERICAS INC

4001453.0500 4001453.1000
N-1155.0500BAEA 529.25 CAD
N-1155.0500BA N-1155.1000BA
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride

Sequence: Tos-Lys-chloromethylketone · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.


Synonym(s): TLCK

Formula: C₁₄H₂₂Cl₂N₂O₃S
Storage Temperature: Freezer
MDL Number: MFCD00065395
CAS Number: 4272-74-6
EINECS: 224-266-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR