Bovine (Tyr34)-Pth (7-34) Amide
Supplier: BACHEM AMERICAS INC
4031202.0500
4031202.1000
N-1110.0500BAEA
774.5
CAD
N-1110.0500BA
N-1110.1000BA
Bovine (Tyr34)-Pth (7-34) Amide
Proteins and Peptides
Sequence: H-Phe-Met-His-Asn-Leu-Gly-Lys-His-Leu-Ser-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Tyr-NH₂
Synonym(s): [Tyr34] Parathyroid Hormone (7-34), amide, bovine
See also the pTH2 receptor agonist TIP39 (H-4878).
An inhibitor of pTH action in vivo.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...