(S)-3-Amino-1-chloro-5-methylhexan-2-one hydrochloride

Supplier: BACHEM AMERICAS INC

4001621.0250 4001621.1000
N-1105.0250BAEA 439.41 CAD
N-1105.0250BA N-1105.1000BA
(S)-3-Amino-1-chloro-5-methylhexan-2-one hydrochloride
(S)-3-Amino-1-chloro-5-methylhexan-2-one hydrochloride

Sequence: H-Leu-chloromethylketone · HCl


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).
Commonly used protease inhibitors as bestatin and the E-64s are derived from leucine. See also H-Leu-NH₂ (E-2650) and Z-Leu-ONp (C-2120).


Leu-CMK, a competitive inhibitor of aminopeptidases and an inhibitor of cathepsin H.

Formula: C₇H₁₄ClNO·HCl
Storage Temperature: Freezer
MDL Number: MFCD00238011
CAS Number: 54518-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR