ZPCK

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Phe-chloromethylketone

4007012.0001 4007012.0005
N-1035.0001BAEA 206.51 CAD
N-1035.0001BA N-1035.0005BA
ZPCK
ZPCK

Sequence: Z-Phe-chloromethylketone


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
See also Z-Phe-OH (C-2310) and Z-Phe-ONp (C-2345).


Synonym(s): ZPCK

Formula: C₁₈H₁₈ClNO₃
Storage Temperature: Freezer
MDL Number: MFCD00037114
CAS Number: 26049-94-5
EINECS: 247-432-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR