Z-Gly-Gly-Phe-Chloromethylketone

Supplier: BACHEM AMERICAS INC

4009487.0025 4009487.0100
N-1030.0025BAEA 877.75 CAD
N-1030.0025BA N-1030.0100BA
Z-Gly-Gly-Phe-Chloromethylketone
Z-Gly-Gly-Phe-Chloromethylketone

Sequence: Z-Gly-Gly-Phe-chloromethylketone


Peptide and amino acid chloromethyl ketones (CMK's) are irreversible inhibitors of cysteine proteases, but they also react with serine proteases. For our amino acid CMK's (e.g. TPCK, TLCK) see the corresponding product family in the 'Amino acid' section.


Z-GGF-CMK, a site-specific inhibitor of chymotrypsin.

Formula: C₂₂H₂₄ClN₃O₅
Storage Temperature: Freezer
MDL Number: MFCD00057806
CAS Number: 35172-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR