Mca-Pro-Leu-Gly-Leu-Glu-Glu-Ala-Dap(Dnp)-NH₂ Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4053570.0001
4053570.0005
M-2670.0001BAEA
645.43
CAD
M-2670.0001BA
M-2670.0005BA
Mca-Pro-Leu-Gly-Leu-Glu-Glu-Ala-Dap(Dnp)-NH₂ Trifluoroacetate
Proteins and Peptides
Highly selective MMP-12 FRET substrate with a kcat/Km value of 1.85 · 10⁵ M⁻¹s⁻¹. Poor substrate of other MMPs with the exception of MMP-13 (kcat/Km = 0.53 · 10 ⁵ M⁻¹s⁻¹) and MMP-9 (0.33 · 10⁵ M⁻¹s⁻¹).