Abz-Phe-Arg-Lys(Dnp)-Pro-OH

Supplier: BACHEM AMERICAS INC

4043877.0001 4043877.0005
M-2590.0001BAEA 309.66 CAD
M-2590.0001BA M-2590.0005BA
Abz-Phe-Arg-Lys(Dnp)-Pro-OH
Proteins and Peptides

Excellent angiotensin I-converting enzyme (ACE) FRET substrate with a Km value of 4.0 µM and a kcat value of 210 s⁻¹. Alves et al. developed a continuous fluorescent assay using Abz-FRK(Dnp)P for the determination of ACE activity in plasma and tissue.

Order Now

SPECIFICATIONS

Protein/peptide name Abz-Phe-Arg-Lys(Dnp)-Pro-OH
Environmentally Preferable
Species
Source
CAS No. 500799-61-1
MDL No. MFCD18782567
Sequence Abz-Phe-Arg-Lys(Dnp)-Pro-OH
Molecular weight 831.89
Molecular formula C₃₉H₄₉N₁₁O₁₀
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR