DABCYL-Lys-HCoV-SARS Replicase Polyprotein 1ab (3235-3246)-Glu-EDANS Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4045664.0001
4045664.0005
M-2575.0001BAEA
1161.77
CAD
M-2575.0001BA
M-2575.0005BA
DABCYL-Lys-HCoV-SARS Replicase Polyprotein 1ab (3235-3246)-Glu-EDANS Trifluoroacetate
Proteins and Peptides
Sensitive internally quenched fluorogenic (FRET) substrate for SARS main protease with a Km value of 17 µM and a kcat value of 1.9 s⁻¹.