Abz-(Asn⁶⁷⁰,Leu⁶⁷¹)-Amyloid beta/A4 Protein Precursor₇₇₀ (669-674)-EDDnp Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4045326.0001 4045326.0005
M-2565.0001BAEA 344.23 CAD
M-2565.0001BA M-2565.0005BA
Abz-(Asn⁶⁷⁰,Leu⁶⁷¹)-Amyloid beta/A4 Protein Precursor₇₇₀ (669-674)-EDDnp Trifluoroacetate
Proteins and Peptides
Intramolecularly quenched fluorescent substrate containing the ortho-aminobenzoyl (Abz) / N-(2,4-dinitrophenyl)ethylenediamine (EDDnp) groups as the donor / acceptor pair. It corresponds to the Swedish-mutated (JMV2236) β-amyloid precursor protein (βAPP) sequence targeted by β-secretase BACE (β-site APP-cleaving activity). This FRET substrate is more selectively cleaved by BACE1 and BACE2 than by cathepsin D, a disintegrin and metalloprotease 10 (ADAM10), tumor necrosis α-converting enzyme (TACE), presenilin-1 (PS1), or presenilin-2 (PS2).