AAF-Arg-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
N-(4-Methoxyphenylazoformyl)-Arg-OH·HCl
4041524.0025
4041524.0100
M-2525.0025BAEA
516.15
CAD
M-2525.0025BA
M-2525.0100BA
AAF-Arg-OH
AAF-Arg-OH
Sequence: N-(4-Methoxyphenylazoformyl)-Arg-OH · HCl
Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).
Synonym(s): Aaf-Arg-OH
Formula:
C₁₄H₂₁ClN₆O₄ MW: 372.81 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD08458656 CAS Number: 442158-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...