Arg-Glu(EDANS)-(Asn⁶⁷⁰,Leu⁶⁷¹)-Amyloid beta/A4 Protein Precursor₇₇₀ (668-675)-Lys(DABCYL)-Arg Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4033536.0001
4033536.0005
M-2470.0001BAEA
1032.67
CAD
M-2470.0001BA
M-2470.0005BA
Arg-Glu(EDANS)-(Asn⁶⁷⁰,Leu⁶⁷¹)-Amyloid beta/A4 Protein Precursor₇₇₀ (668-675)-Lys(DABCYL)-Arg Trifluoroacetate
Proteins and Peptides
Fluorogenic (FRET) substrate for pro-memapsin-2 containing the β-secretase site EVNLDAEF of the Swedish mutation of APP. The kinetic parameters at pH 4.5 are Km = 5.4 µM and kcat = 0.24 min⁻¹.