FITC-Tyr-Val-Ala-Asp-Ala-Pro-Lys(Dnp)-OH (Contains FITC isomer I)
Supplier: BACHEM AMERICAS INC
4027937.0001 4027937.0005
M-2285.0001BAEA 697.9 CAD
M-2285.0001BA M-2285.0005BA
FITC-Tyr-Val-Ala-Asp-Ala-Pro-Lys(Dnp)-OH (Contains FITC isomer I)
Proteins and Peptides
FITC-YVADAPK(Dnp) is a specific FRET substrate for the determination of caspase-1 and caspase-1 like enzyme activities. Cleavage of FITC-YVADAPK(Dnp) at the P1 Asp residue results in a continuous fluorescent assay. It is useful both in FACS and fluorescence microscopy experiments. Caspase-3 is only weakly active using this substrate. See also our reference substance M-2280.