Mca-Pro-Leu-Gly-Pro-D-Lys(Dnp)-OH
Supplier: BACHEM AMERICAS INC
4027687.0001
4027687.0005
M-2270.0001BAEA
490.51
CAD
M-2270.0001BA
M-2270.0005BA
Mca-Pro-Leu-Gly-Pro-D-Lys(Dnp)-OH
Proteins and Peptides
See also M-2460 and M-2465.
Specific highly fluorescent FRET substrate for the determination of the thimet oligopeptidase (EP 24.15, EC 3.4.24.15.) activity. See also M-1975, Mca-Pro-Leu-OH.