Human Mca-(endo-1a-Dap(Dnp))-TNF-alpha (-5 to +6) amide Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4031302.0001
4031302.0005
M-2255.0001BAEA
645.43
CAD
M-2255.0001BA
M-2255.0005BA
Human Mca-(endo-1a-Dap(Dnp))-TNF-alpha (-5 to +6) amide Trifluoroacetate
Proteins and Peptides
ADAM-17 endopeptidase is also called TACE or TNFα-converting enzyme.
Mca-PLAQAVDpaRSSSR-amide, a fluorogenic (FRET) peptide substrate for tumor necrosis factor (TNF) converting enzyme (TACE, ADAM-17 endopeptidase).