NFF 3

Supplier: BACHEM AMERICAS INC

4030842.0001 4030842.0005
M-2110.0001BAEA 722.85 CAD
M-2110.0001BA M-2110.0005BA
NFF 3
NFF 3

Sequence: Mca-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(Dnp)-NH₂
Synonym(s): NFF-3


This FRET substrate Mca-RPKPVE-Nval-WRK(Dnp) amide was hydrolyzed rapidly by stromelysin 1 (MMP-3) with kcat/Km= 218000 M⁻¹s⁻¹ and very slowly by gelatinase B (MMP-9) (kcat/Km= 10100 M⁻¹s⁻¹). There was no hydrolysis by MMP-1 and MMP-2. For a fluorescent standard see M-2250.

Formula: C₇₈H₁₁₀N₂₂O₂₀
Storage Temperature: Freezer
MDL Number: MFCD03844467
CAS Number: 158584-09-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR