DABCYL-gamma-Abu-Ser-Gln-Asn-Tyr-Pro-Ile-Val-Gln-EDANS Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030716.0001 4030716.0005
M-1865.0001BAEA 774.5 CAD
M-1865.0001BA M-1865.0005BA
DABCYL-gamma-Abu-Ser-Gln-Asn-Tyr-Pro-Ile-Val-Gln-EDANS Trifluoroacetate
Proteins and Peptides
This fluorogenic HIV-1 protease substrate consists of an octapeptide with a fluorescent donor (EDANS) and a quenching acceptor (DABCYL), attached at the COOH- and NH₂-termini. The γ-Abu spacer was inserted to avoid potential steric hindrance of substrate binding by the bulky acceptor. This FRET substrate is cleaved by the HIV-1 protease at the Tyr-Pro bond which results in a time-dependent increase in fluorescence intensity; excitation at 340 nm, emission at 490 nm.