FA-Ala-Arg-OH
Supplier: BACHEM AMERICAS INC
4013709.0050 4013709.0250
M-1810.0050BAEA 450.44 CAD
M-1810.0050BA M-1810.0250BA
FA-Ala-Arg-OH
Proteins and Peptides
The FA-peptides M-1400 and M-1410 are common ACE substrates and thus filed in the corresponding product family. For our 3-(2-furyl)acryloyl-amino acids (FA-amino acids) and the labeling reagent FA-OSu (Q-1245), please see the Amino Acids section.
Substrate for human plasma carboxypeptidase N and membrane-bound carboxypeptidase D.