Pro-Leu-Gly hydroxamate hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Pro-Leu-Gly-NHOH·HCl

4012165.0050 4012165.0250
M-1775.0050BAEA 283.94 CAD
M-1775.0050BA M-1775.0250BA
Pro-Leu-Gly hydroxamate hydrochloride
Pro-Leu-Gly hydroxamate hydrochloride

Sequence: H-Pro-Leu-Gly-NHOH · HCl


Affinity ligand for the purification of human collagenases. Inhibitor of MMP-8, Ki 19 μM (Grams et al., 1995).

Formula: C₁₃H₂₅ClN₄O₄
MW: 336.817 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00238157
CAS Number: 120928-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR