Suc-FLF-thiobenzyl ester
Supplier: BACHEM AMERICAS INC
Synonyms:
Suc-Phe-Leu-Phe-SBzl
4006633.0005
4006633.0025
M-1740.0005BAEA
209
CAD
M-1740.0005BA
M-1740.0025BA
Suc-FLF-thiobenzyl ester
Suc-FLF-thiobenzyl ester
Sequence: Suc-Phe-Leu-Phe-SBzl
Suc-FLF-thiobenzyl ester, a very sensitive substrate for the determination of cathepsin G activity. It is also cleaved by chymotrypsin, chymotrypsin-like serine proteases from skin as well as rat mast cell proteases.
Formula:
C₃₅H₄₁N₃O₆S MW: 631.788 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00077164 CAS Number: 80651-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...