Pz-Pro-Leu-Gly-Pro-D-Arg-OH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4000833.0250
4000833.1000
M-1715.0250BAEA
413.58
CAD
M-1715.0250BA
M-1715.1000BA
Pz-Pro-Leu-Gly-Pro-D-Arg-OH Trifluoroacetate
Proteins and Peptides
See also M-2460 and M-2465.
Chromogenic substrate for the assay of collagenase and thimet oligopeptidase (Pz-peptidase, metalloendopeptidase 24.15, collagenase-like peptidase). Enzymatic cleavage yields Pz-Pro-Leu-OH, G-2970 (or Pz-Pro-OH, E-2275). The cleavage can be monitored by analytical HPLC, detection at 320 nm.