AM001287

Supplier: BACHEM AMERICAS INC

4006749.0025 4006749.0100
M-1540.0025BAEA 645.43 CAD
M-1540.0025BA M-1540.0100BA
AM001287
AM001287

Sequence: H-Leu-Ser-p-nitro-Phe-Nle-Ala-Leu-OMe
Synonym(s): LSF(NO2)-Nle-AL-Methyl Ester


Standard substrate for chymosin activity determination. This peptide is also a good substrate for pepsin, penicillopepsin, and gastricsin.

Formula: C₃₆H₅₆F₃N₇O₁₂
Storage Temperature: Freezer
MDL Number: MFCD00211055
CAS Number: 99764-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR