Ohip-his-leu

Supplier: BACHEM AMERICAS INC

Synonyms: p-Hydroxyhippuryl-His-Leu-OH

4004645.0250 4004645.1000
M-1505.0250BAEA 903.62 CAD
M-1505.0250BA M-1505.1000BA
Ohip-his-leu
Ohip-his-leu

Sequence: p-Hydroxyhippuryl-His-Leu-OH


Substrate for a simple, accurate, and reproducible colorimetric determination of angiotensin I-converting enzyme (ACE) activity.

Formula: C₂₁H₂₇N₅O₆
Storage Temperature: Freezer
MDL Number: MFCD00038808
CAS Number: 77697-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR