Hippuryl-His-Leu-OH
Supplier: BACHEM AMERICAS INC
4002147.0250 4002147.1000
M-1485.0250BAEA 206.51 CAD
M-1485.0250BA M-1485.1000BA
Hippuryl-His-Leu-OH
Proteins and Peptides
Substrate for a spectrophotometric assay of angiotensin I-converting enzyme. ACE cleavage of Hip-His-Leu yields hippuric acid and His-Leu (G-2310), which can be quantified by generating a fluorescent adduct with o-phthalaldehyde. Schwager et al. developed a high-throughput fluorometric assay for ACE employing Hip-His-Leu.