FA-Leu-Gly-Pro-Ala-OH
Supplier: BACHEM AMERICAS INC
4006713.0025 4006713.0100
M-1385.0025BAEA 464.74 CAD
M-1385.0025BA M-1385.0100BA
FA-Leu-Gly-Pro-Ala-OH
Proteins and Peptides
The FA-peptides M-1400 and M-1410 are common ACE substrates and thus filed in the corresponding product family. For our 3-(2-furyl)acryloyl-amino acids (FA-amino acids) and the labeling reagent FA-OSu (Q-1245), please see the Amino Acids section.
Substrate for a versatile, convenient, continuously recording spectrophotometric assay of collagenase.