Ac-Lys(Ac)-D-Ala-D-Ala-OH
Supplier: BACHEM AMERICAS INC
4033018.0050 4033018.0250
M-1325.0050BAEA 529.09 CAD
M-1325.0050BA M-1325.0250BA
Ac-Lys(Ac)-D-Ala-D-Ala-OH
Proteins and Peptides
See also F-3260.
Diacetyl-Lys-D-Ala-D-Ala (DALAA) is a substrate for penicillin-sensitive D-alanine carboxypeptidases (DD-carboxypeptidases).