Z-Phe-His-Leu-OH

Supplier: BACHEM AMERICAS INC

4000599.0250 4000599.1000
M-1305.0250BAEA 464.74 CAD
M-1305.0250BA M-1305.1000BA
Z-Phe-His-Leu-OH
Z-Phe-His-Leu-OH

Sequence: Z-Phe-His-Leu-OH


Substrate for angiotensin I-converting enzyme. Cleavage of Z-FHL by ACE yields His-Leu (G-2310), which can be quantified by generating a fluorescent adduct with o-phthalaldehyde.

Formula: C₂₉H₃₅N₅O₆
Storage Temperature: Freezer
MDL Number: MFCD00155532
CAS Number: 28458-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR