Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: Z-L-Lys-SBzl hydrochloride, BLT HCl, Z-Lys-SBzl·HCl, (S)-S-Benzyl 6-amino-2-(((benzyloxy)carbonyl)amino)hexanethioate hydrochloride

4002677.0050 4002677.0250 4002677.1000
M-1300.0050BAEA 192.25 CAD
M-1300.0050BA M-1300.0250BA M-1300.1000BA
Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride
Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride

Sequence: Z-Lys-SBzl · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.


Substrate for a sensitive colorimetric assay for trypsin-like enzymes, e.g. kallikreins, thrombin, plasmin, trypsin, and enteropeptidase. A sensitive, coupled assay for plasminogen activators (urokinase and tPA) using this substrate has been described.

Formula: C₂₁H₂₇ClN₂O₃S
MW: 422.968 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058089
CAS Number: 69861-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR