Z-Glu-Tyr-OH
Supplier: BACHEM AMERICAS INC
4001669.0005
4001669.0025
M-1245.0005BAEA
309.81
CAD
M-1245.0005BA
M-1245.0025BA
Z-Glu-Tyr-OH
Z-Glu-Tyr-OH
Sequence: Z-Glu-Tyr-OH
Substrate for porcine cathepsin A and acid carboxypeptidases from A. niger.and A. kawachii.
Formula:
C₂₂H₂₄N₂O₈ MW: 444.44 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00037189 CAS Number: 988-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...