Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

4002371.0025 4002371.0100
M-1130.0025BAEA 284.04 CAD
M-1130.0025BA M-1130.0100BA
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride

Sequence: Bz-Arg-OEt · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Synonym(s): L-BAEE

Formula: C₁₅H₂₂N₄O₃
Storage Temperature: Freezer
MDL Number: MFCD00012579
CAS Number: 2645-08-1
EINECS: 220-157-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR