Abz-Gly-p-nitro-Phe-Pro-OH Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4003531.0050 4003531.0250
M-1100.0050BAEA 926.18 CAD
M-1100.0050BA M-1100.0250BA
Abz-Gly-p-nitro-Phe-Pro-OH Trifluoroacetate
Proteins and Peptides
Fluorogenic (FRET) substrate for angiotensin I-converting enzyme (ACE). Enzymatic cleavage of M-1100 yields Abz-Gly-OH (E-2920) emitting at 420 nm.