H-Ala-Ala-Ala-OMe Acetate
Supplier: BACHEM AMERICAS INC
4010702.0001
4010702.0005
M-1090.0001BAEA
697.9
CAD
M-1090.0001BA
M-1090.0005BA
H-Ala-Ala-Ala-OMe Acetate
Proteins and Peptides
See also I-1270, L-1000, L-1090, L-1180, L-1335, L-1385, L-1400, M-1170, and M-1250.
AAA-OMe, substrate for a simultaneous determination of the esterase and the peptidase activities of pancreatic elastase.