APNE

Supplier: BACHEM AMERICAS INC

4000453.0001 4000453.0005
M-1055.0001BAEA 671.97 CAD
M-1055.0001BA M-1055.0005BA
APNE
APNE

Sequence: Ac-DL-Phe-β-naphthyl ester


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
See also Z-Phe-OH (C-2310) and Z-Phe-ONp (C-2345).


Synonym(s): DL-APNE

Formula: C₂₁H₁₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00037746
CAS Number: 20874-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR