Ac-L-Lys-OMe·HCI

Supplier: BACHEM AMERICAS INC

4012001.0001 4012001.0005 4012001.0025
M-1045.0001BAEA 209 CAD
M-1045.0001BA M-1045.0005BA M-1045.0025BA
Ac-L-Lys-OMe·HCI
Ac-L-Lys-OMe·HCI

Sequence: Ac-Lys-OMe · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.


Synonym(s): L-ALME

Formula: C₉H₁₈N₂O₃·HCl
Storage Temperature: Freezer
MDL Number: MFCD00003570
CAS Number: 20911-93-7
EINECS: 244-111-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR