Acetyltrialanine

Supplier: BACHEM AMERICAS INC

Synonyms: Ac-Ala-Ala-Ala-OH

4003070.0250
M-1010.1000BAEA 2008.97 CAD
M-1010.1000BA M-1010.0250BA
Acetyltrialanine
Acetyltrialanine

Sequence: Ac-Ala-Ala-Ala-OH


Ac-AAA, suitable substrate for acylaminoacyl peptidase (N-acyl-peptide hydrolase). This enzyme releases acylamino acids from N-acylated peptides and may therefore play a role in protein degradation.

Formula: C₁₁H₁₉N₃O₅
MW: 273.289 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00002642
CAS Number: 19245-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR