Boc-Gln-Ala-Arg-pNA
Supplier: BACHEM AMERICAS INC
4037278.0050 4037278.0250
L-2140.0050BAEA 681.75 CAD
L-2140.0050BA L-2140.0250BA
Boc-Gln-Ala-Arg-pNA
Proteins and Peptides
Boc-QAR-pNA, chromogenic substrate for trypsin and matriptase-2.