Z-Phe-Leu-Glu-pNA

Supplier: BACHEM AMERICAS INC

4037068.0025 4037068.0100
L-2135.0025BAEA 794.35 CAD
L-2135.0025BA L-2135.0100BA
Z-Phe-Leu-Glu-pNA
Z-Phe-Leu-Glu-pNA

Sequence: Z-Phe-Leu-Glu-pNA


Z-FLE-pNA, a chromogenic substrate for glutamyl endopeptidases.

Formula: C₃₄H₃₉N₅O₉
MW: 661.712 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01940686
CAS Number: 104634-10-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR