H-D-Pro-Phe-Arg-pNA·2·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-D-Pro-Phe-Arg-pNA·2 HCl

4029234.0025 4029234.0100
L-2120.0025BAEA 1221.34 CAD
L-2120.0025BA L-2120.0100BA
H-D-Pro-Phe-Arg-pNA·2·HCl
H-D-Pro-Phe-Arg-pNA·2·HCl

Sequence: H-D-Pro-Phe-Arg-pNA · 2 HCl


See also I-1160, I-1960, L-1545, M-1130, and M-1140.


pFR-pNA, chromogenic substrate for the determination of plasma kallikrein-like activity. CAS Number (net): 64816-19-9.

Formula: C₂₆H₃₆Cl₂N₈O₅
MW: 611.525 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00274447
CAS Number: 62354-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR