Z-D-Arg-Gly-Arg-pNA·2 HCl
Supplier: BACHEM AMERICAS INC
4029223.0025
4029223.0100
L-2115.0025BAEA
656.14
CAD
L-2115.0025BA
L-2115.0100BA
Z-D-Arg-Gly-Arg-pNA·2 HCl
Proteins and Peptides
Z-RGR-pNA is a chromogenic substrate for the determination of factor Xa.