Suc-Ala-Pro-pNA
Supplier: BACHEM AMERICAS INC
4028677.0050
4028677.0250
L-2090.0050BAEA
903.62
CAD
L-2090.0050BA
L-2090.0250BA
Suc-Ala-Pro-pNA
Proteins and Peptides
See also I-1350 and K-1490.
Suc-AP-pNA, chromogenic substrate for prolyl endopeptidase.