H-Lys-Ala-pNA · 2 HCl
Supplier: BACHEM AMERICAS INC
4028676.0050 4028676.0250
L-2085.0050BAEA 439.41 CAD
L-2085.0050BA L-2085.0250BA
H-Lys-Ala-pNA · 2 HCl
Proteins and Peptides
Chromogenic substrate for dipeptidyl aminopeptidase II (DPPII), also cleaved by a dipeptidyl peptidase V (DPP V) from Aspergillus fumigatus.