H-Trp-Phe-Tyr-Ser(PO₃H₂)-Pro-Arg-pNA Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4028474.0001 4028474.0005
L-2075.0001BAEA 529.57 CAD
L-2075.0001BA L-2075.0005BA
H-Trp-Phe-Tyr-Ser(PO₃H₂)-Pro-Arg-pNA Trifluoroacetate
Proteins and Peptides
WFYpSPR-pNA, chromogenic substrate for Pin1, an essential and conserved mitotic peptidyl-prolyl isomerase (PPIase), that specifically recognizes the phosphoserine-proline bonds present in mitotic phosphoproteins (kcat/Km = 20160 mM⁻¹s⁻¹).